3,6-Dimethoxy-9H-carbazole 97%

Supplier: Ambeed
Warning

A191775-100MG A191775-250MG
103816-402EA 24 USD
103816-402 103847-004 77236-926 77236-928
3,6-Dimethoxy-9H-carbazole 97%
3,6-Dimethoxy-9H-carbazole

Precaución: for Research Use Only

Formula: C₁₄H₁₃NO₂
Storage Temperature: Ambient
MDL Number: MFCD00626046
CAS Number: 57103-01-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR