3,6-Diiodocarbazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,6-Diiodo-9H-carbazole
A121431-10G
A121431-25G
A121431-500G
A121431-1G
A121431-5G
103800-072EA
8
USD
103800-072
77161-322
77161-324
77161-326
77297-830
77312-194
3,6-Diiodocarbazole 98%
3,6-Diiodocarbazole
Precaución: for Research Use Only
Formula:
C₁₂H₇I₂N Boiling Pt: 498.3 °C (760 mm Hg) Melting Pt: 211…212 °C Flash Pt: 255.2 °C Storage Temperature: Ambient |
MDL Number:
MFCD00450017 CAS Number: 57103-02-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...