3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dihydro-2H-pyran-4-boronic acid Pinacol ester, 2-(3,6-Dihydro-2H-pyran-4-yl)-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, 3,6-Dihydro-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2H-pyran, 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)-3,6-dihydro-2H-pyran

A249729-250MG A249729-100G A249729-10G
103811-886EA 20 USD
103811-886 103851-724 77031-476 77568-426 77593-576
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester 98%
3,6-Dihydro-2H-pyran-4-boronic acid pinacol ester

Precaución: For research use only.

Formula: C₁₁H₁₉BO₃
MW: 210.08 g/mol
Boiling Pt: 238.6 °C (760 mm Hg)
Melting Pt: 63…67 °C
Flash Pt: 98.124 °C
Storage Temperature: Freezer
MDL Number: MFCD11052631
CAS Number: 287944-16-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR