3,6-Difluoropyridin-2-amine 95%

Supplier: Ambeed
Warning

A146454-100MG A146454-250MG
103803-668EA 195 USD
103803-668 103841-760 77010-556 77010-558
3,6-Difluoropyridin-2-amine 95%
3,6-Difluoropyridin-2-amine

Precaución: for Research Use Only

Formula: C₅H₄F₂N₂
Boiling Pt: 190.0±35.0 °C (760 mm Hg)
Flash Pt: 68.7 °C
Storage Temperature: Ambient
MDL Number: MFCD13175493
CAS Number: 944799-22-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR