3,6-Difluoro-2-methylbenzaldehyde 95%
Supplier: Ambeed
|
Warning
|
A597372-1G
A597372-100MG
103828-962EA
217
USD
103828-962
76844-250
77103-988
3,6-Difluoro-2-methylbenzaldehyde 95%
3,6-Difluoro-2-methylbenzaldehyde
Precaución: for Research Use Only
Formula:
C₈H₆F₂O MW: 156.129 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD16250132 CAS Number: 1378525-21-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...