3,6-Dichloropyridazine-4-carbonitrile 98%

Supplier: Ambeed
Warning

A133746-250MG A133746-1G A133746-25G
103801-912EA 42 USD
103801-912 76972-240 77314-598 77881-062
3,6-Dichloropyridazine-4-carbonitrile 98%
3,6-Dichloropyridazine-4-carbonitrile

Precaución: for Research Use Only

Formula: C₅HCl₂N₃
Storage Temperature: Ambient
MDL Number: MFCD16621820
CAS Number: 35857-93-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR