2-Cyano-3,6-dichloropyridine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,6-Dichloropicolinonitrile
A196397-10G
A196397-5G
103816-842EA
23
USD
103816-842
103847-454
76825-390
77302-518
2-Cyano-3,6-dichloropyridine 95%
2-Cyano-3,6-dichloropyridine
Precaución: for Research Use Only
Formula:
C₆H₂Cl₂N₂ MW: 173 g/mol Boiling Pt: 274.5 °C (760 mm Hg) Melting Pt: 96…98 °C Flash Pt: 119.8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00546824 CAS Number: 1702-18-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...