2,5-Dichloropyridine-6-carboxaldehyde 95%

Supplier: Ambeed
Warning

Synonyms: 3,6-Dichloropicolinaldehyde

A347127-100MG A347127-1G A347127-5G
103825-954EA 35 USD
103825-954 103836-944 76788-340 76840-016 76840-018
2,5-Dichloropyridine-6-carboxaldehyde 95%
2,5-Dichloropyridine-6-carboxaldehyde

Precaución: for Research Use Only

Formula: C₆H₃Cl₂NO
MW: 176 g/mol
Boiling Pt: 234.2 °C (760 mm Hg)
Flash Pt: 95 °C
Storage Temperature: Refrigerator
MDL Number: MFCD12963768
CAS Number: 343781-53-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR