3,6-Dichloro-4-methylpyridazine 98%
Supplier: Ambeed
|
Warning
|
A180732-100G
A180732-10G
A180732-500G
A180732-25G
A180732-5G
103845-914EA
7
USD
103845-914
77096-822
77096-824
77096-826
77301-690
77301-692
3,6-Dichloro-4-methylpyridazine 98%
3,6-Dichloro-4-methylpyridazine
Precaución: for Research Use Only
Formula:
C₅H₄Cl₂N₂ Boiling Pt: 272.1 °C (760 mm Hg) Melting Pt: 84…89 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006465 CAS Number: 19064-64-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...