3,6-Dichloro-4-methoxypyridazine 97%

Supplier: Ambeed
Warning

A279814-10G A279814-250MG A279814-1G A279814-25G A279814-5G
103813-488EA 7 USD
103813-488 103853-432 77032-144 77032-146 77093-354 77093-356 77093-358
3,6-Dichloro-4-methoxypyridazine 97%
3,6-Dichloro-4-methoxypyridazine

Precaución: for Research Use Only

Formula: C₅H₄Cl₂N₂O
MW: 179.0000 g/mol
Melting Pt: 130…134 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00477252
CAS Number: 70952-62-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR