3,6-Dichloro-4,5-dimethylpyridazine 98%

Supplier: Ambeed
Warning

A126204-100G A126204-10G A126204-1G A126204-25G A126204-5G
103800-776EA 14 USD
103800-776 77271-798 77271-800 77298-108 77312-964 77312-966
3,6-Dichloro-4,5-dimethylpyridazine 98%
3,6-Dichloro-4,5-dimethylpyridazine

Precaución: for Research Use Only

Formula: C₆H₆Cl₂N₂
MW: 177.031 g/mol
Boiling Pt: 317.1 °C (760 mm Hg)
Storage Temperature: Refrigerator
MDL Number: MFCD00067745
CAS Number: 34584-69-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR