3,6-Dibromopyridazine 97%
Supplier: Ambeed
|
Warning
|
A146562-100G
A146562-10G
A146562-1G
A146562-25G
A146562-5G
A146562-500G
103803-680EA
5
USD
103803-680
103841-772
77110-064
77110-066
77317-100
77317-102
77317-104
77611-350
3,6-Dibromopyridazine 97%
3,6-Dibromopyridazine
Precaución: for Research Use Only
Formula:
C₄H₂Br₂N₂ Boiling Pt: 327.5 °C (760 mm Hg) Flash Pt: 151.862 °C Storage Temperature: Ambient |
MDL Number:
MFCD00233947 CAS Number: 17973-86-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...