3,6-Dibromophenanthrene-9,10-dione 97%
Supplier: Ambeed
|
Warning
|
A149294-100G
A149294-10G
A149294-250MG
A149294-500G
A149294-25G
A149294-5G
A149294-1G
103804-024EA
7
USD
103804-024
103842-142
77272-914
77272-916
77272-918
77272-920
77299-718
77299-720
77317-628
3,6-Dibromophenanthrene-9,10-dione 97%
3,6-Dibromophenanthrene-9,10-dione
Precaución: for Research Use Only
Formula:
C₁₄H₆Br₂O₂ Boiling Pt: 501 °C (760 mm Hg) Flash Pt: 177.5 °C Storage Temperature: Ambient |
MDL Number:
MFCD00451676 CAS Number: 53348-05-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...