3,6-Dibromobenzene-1,2-diamine 97%

Supplier: Ambeed
Warning

A189966-10G A189966-250MG A189966-1G A189966-25G A189966-5G
103816-244EA 10 USD
103816-244 103846-838 76986-048 76986-050 77084-624 77084-626 77084-628
3,6-Dibromobenzene-1,2-diamine 97%
3,6-Dibromobenzene-1,2-diamine

Precaución: for Research Use Only

Formula: C₆H₆Br₂N₂
MW: 265.93 g/mol
Boiling Pt: 320.9±37.0 °C (760 mm Hg)
Melting Pt: 92…94 °C
Flash Pt: 147.9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD19288763
CAS Number: 69272-50-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR