3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 97%

Supplier: Ambeed
Warning

A130224-100G A130224-10G A130224-250MG A130224-1G A130224-5G
103801-400EA 6 USD
103801-400 77271-970 77271-972 77271-974 77313-828 77313-830
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine 97%
3,6-Di(2-pyridyl)-1,2,4,5-tetrazine

Precaución: for Research Use Only

Formula: C₁₂H₈N₆
Boiling Pt: 509.1 °C (760 mm Hg)
Melting Pt: 225 °C
Flash Pt: 245.7 °C
Storage Temperature: Ambient
MDL Number: MFCD00121717
CAS Number: 1671-87-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR