3,6-Bis(3,5-dimethyl-1H-pyrazol-1-yl)-1,2,4,5-tetrazine 97%
Supplier: Ambeed
|
Danger
|
A300012-100G
A300012-10G
A300012-1G
A300012-25G
103814-252EA
6
USD
103814-252
103835-402
77239-238
77239-240
77305-782
77305-784
3,6-Bis(3,5-dimethyl-1H-pyrazol-1-yl)-1,2,4,5-tetrazine 97%
3,6-Bis(3,5-dimethyl-1H-pyrazol-1-yl)-1,2,4,5-tetrazine
Precaución: for Research Use Only
Formula:
C₁₂H₁₄N₈ MW: 270.29 g/mol Boiling Pt: 533.7 °C (760 mm Hg) Melting Pt: 216…218 °C Flash Pt: 276.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00173232 CAS Number: 30169-25-6 UN: 1325 ADR: 4.1,Ⅱ |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...