1,14-Diamino-3,6,9,12-tetraoxatetradecane 95%

Supplier: Ambeed
Warning

Synonyms: Triethylene Glycol bis(2-aminoethyl) ether, 3,6,9,12-Tetraoxatetradecane-1,14-diamine

A160035-100MG A160035-250MG A160035-1G A160035-5G
103805-320EA 124 USD
103805-320 103843-550 77138-094 77138-096 77319-458 77319-460
1,14-Diamino-3,6,9,12-tetraoxatetradecane 95%
1,14-Diamino-3,6,9,12-tetraoxatetradecane

Precaución: for Research Use Only

Formula: C₁₀H₂₄N₂O₄
MW: 236.31 g/mol
Boiling Pt: 167…168 °C (3 mm Hg)
Flash Pt: 157.3 °C
Storage Temperature: Refrigerator
MDL Number: MFCD01365938
CAS Number: 68960-97-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR