3,6,6-Trimethyl-6,7-dihydro-1H-indol-4(5H)-one 95%

Supplier: Ambeed
Warning

A281123-250MG A281123-1G
103813-538EA 65 USD
103813-538 103853-490 76827-560 77305-384
3,6,6-Trimethyl-6,7-dihydro-1H-indol-4(5H)-one 95%
3,6,6-Trimethyl-6,7-dihydro-1H-indol-4(5H)-one

Precaución: for Research Use Only

Formula: C₁₁H₁₅NO
MW: 177.24 g/mol
Storage Temperature: Ambient
MDL Number: MFCD08060464
CAS Number: 56008-20-9

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR