3,5-Di-tert-butylpyrocatechol 95%

Supplier: Ambeed
Warning

Synonyms: 3,5-Di-tert-butylcatechol

A296117-25G A296117-100G A296117-5G
103835-220EA 9 USD
103835-220 76837-886 77277-252 77305-682
3,5-Di-tert-butylpyrocatechol 95%
3,5-Di-tert-butylpyrocatechol

Precaución: for Research Use Only

Formula: C₁₄H₂₂O₂
MW: 222.33 g/mol
Melting Pt: 95…100 °C
Storage Temperature: Ambient
MDL Number: MFCD00008819
CAS Number: 1020-31-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR