3,5-Di-tert-butylbenzoic acid 99%

Supplier: Ambeed
Warning

A253209-100G A253209-10G A253209-250MG A253209-1G A253209-25G A253209-5G
103812-084EA 7 USD
103812-084 103851-932 77031-598 77031-600 77031-602 77091-354 77091-356 77091-358
3,5-Di-tert-butylbenzoic acid 99%
3,5-Di-tert-butylbenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₂₂O₂
MW: 234.34 g/mol
Boiling Pt: 314.3 °C (760 mm Hg)
Melting Pt: 173…175 °C
Flash Pt: 155.1 °C
Storage Temperature: Ambient
MDL Number: MFCD00082727
CAS Number: 16225-26-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR