3,5-Di-tert-butylbenzaldehyde 97%

Supplier: Ambeed
Warning

Synonyms: 3,5-Bis(tert-butyl)benzaldehyde

A107437-100G A107437-10G A107437-250MG A107437-25G A107437-1G A107437-5G
103797-190EA 10 USD
103797-190 103822-960 77025-528 77025-530 77025-532 77025-534 77309-808 77309-810
3,5-Di-tert-butylbenzaldehyde 97%
3,5-Di-tert-butylbenzaldehyde

Precaución: for Research Use Only

Formula: C₁₅H₂₂O
MW: 218.34 g/mol
Boiling Pt: 266.2 °C (760 mm Hg)
Melting Pt: 85…89 °C
Storage Temperature: Ambient
MDL Number: MFCD00026298
CAS Number: 17610-00-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR