3,5-Diphenyl-1H-pyrazole 98%

Supplier: Ambeed
Warning

A171255-100G A171255-10G A171255-25G A171255-5G
103844-888EA 9 USD
103844-888 77273-724 77273-726 77301-110 77301-112
3,5-Diphenyl-1H-pyrazole 98%
3,5-Diphenyl-1H-pyrazole

Precaución: for Research Use Only

Formula: C₁₅H₁₂N₂
MW: 220.27 g/mol
Melting Pt: 199…203 °C
Flash Pt: 207.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00039675
CAS Number: 1145-01-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR