4-Amino-3,5-dimethylpyridine 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethylpyridin-4-amine

A114979-100MG A114979-250MG A114979-5G
103798-510EA 47 USD
103798-510 103824-352 77271-168 77271-170 77311-174
4-Amino-3,5-dimethylpyridine 98%
4-Amino-3,5-dimethylpyridine

Precaución: for Research Use Only

Formula: C₇H₁₀N₂
MW: 122.1700 g/mol
Boiling Pt: 277.801 °C (760 mm Hg)
Melting Pt: 103…107 °C
Flash Pt: 146.292 °C
Storage Temperature: Ambient
MDL Number: MFCD03413657
CAS Number: 43078-60-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR