3,5-Dimethyl-1H-pyrazole-4-carboxylic acid 97%

Supplier: Ambeed
Warning

A423104-1G A423104-25G A423104-5G
103827-430EA 19 USD
103827-430 103838-478 76842-112 76842-114 76842-116
3,5-Dimethyl-1H-pyrazole-4-carboxylic acid 97%
3,5-Dimethyl-1H-pyrazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C6H8N2O2
Melting Pt: 273 °C
Storage Temperature: Ambient
MDL Number: MFCD00159642
CAS Number: 113808-86-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR