3,5-Dimethylpyrazine-2-carboxylic acid 98%

Supplier: Ambeed
Warning

A139907-250MG A139907-5G
103802-794EA 309 USD
103802-794 76811-724 77315-900
3,5-Dimethylpyrazine-2-carboxylic acid 98%
3,5-Dimethylpyrazine-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₇H₈N₂O₂
MW: 152.15 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD18070837
CAS Number: 946493-27-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR