3,5-Dimethylisoxazole-4-boronic acid 95%
Supplier: Ambeed
|
Warning
|
A124422-100G
A124422-10G
A124422-250MG
A124422-25G
A124422-500G
A124422-1G
A124422-5G
103800-530EA
8
USD
103800-530
77285-046
77285-048
77285-050
77285-052
77285-054
77312-666
77312-668
3,5-Dimethylisoxazole-4-boronic acid 95%
3,5-Dimethylisoxazole-4-boronic acid
Precaución: for Research Use Only
Formula:
C₅H₈BNO₃ Boiling Pt: 333.9 °C (760 mm Hg) Melting Pt: 108…113 °C Flash Pt: 155.8 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD02677945 CAS Number: 16114-47-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...