3,5-Dimethylisoxazole-4-carboxylic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethylisoxasole-4-carboxylic acid

A392866-5G A392866-100G A392866-10G A392866-1G
103826-912EA 17 USD
103826-912 103837-952 76841-348 77115-670 77115-672 77307-024
3,5-Dimethylisoxazole-4-carboxylic acid 98%
3,5-Dimethylisoxazole-4-carboxylic acid

Precaución: for Research Use Only

Formula: C₆H₇NO₃
MW: 141.13 g/mol
Boiling Pt: 301.8 °C (760 mm Hg)
Melting Pt: 141…145 °C
Flash Pt: 136.3 °C
Storage Temperature: Ambient
MDL Number: MFCD00051657
CAS Number: 2510-36-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR