3,5-Dimethyl-4-nitropyrazole 98%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Dimethyl-4-nitro-1H-pyrazole
A362346-25G
A362346-5G
A362346-100G
A362346-10G
A362346-1G
A362346-500G
103837-330EA
6
USD
103837-330
76840-542
76840-544
77278-338
77278-340
77278-342
77278-344
3,5-Dimethyl-4-nitropyrazole 98%
3,5-Dimethyl-4-nitropyrazole
Precaución: for Research Use Only
Formula:
C₅H₇N₃O₂ Boiling Pt: 304.5 °C (760 mm Hg) Melting Pt: 123…124 °C Flash Pt: 138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052513 CAS Number: 14531-55-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...