3,5-Dimethyl-4-iodo-1H-pyrazole 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Dimethyl-4-iodopyrazole
A187688-100G
A187688-10G
A187688-1G
A187688-25G
A187688-5G
103846-628EA
6
USD
103846-628
77012-180
77012-182
77012-184
77084-340
77084-342
3,5-Dimethyl-4-iodo-1H-pyrazole 97%
3,5-Dimethyl-4-iodo-1H-pyrazole
Precaución: for Research Use Only
Formula:
C₅H₇IN₂ MW: 222.03 g/mol Boiling Pt: 203.2 °C (760 mm Hg) Melting Pt: 70…72 °C Flash Pt: 133.6 °C Storage Temperature: Ambient |
MDL Number:
MFCD00040247 CAS Number: 2033-45-6 |
Learn more
![](/m-us.vwr.com/en_US/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...