3,5-Dimethyl-4-iodoisoxazole 97%

Supplier: Ambeed
Warning

A369642-100G A369642-10G
103837-466EA 6 USD
103837-466 77015-956 77015-958
3,5-Dimethyl-4-iodoisoxazole 97%
3,5-Dimethyl-4-iodoisoxazole

Precaución: for Research Use Only

Formula: C₅H₆INO
Boiling Pt: 295.4 °C (760 mm Hg)
Storage Temperature: Ambient
MDL Number: MFCD00173743
CAS Number: 10557-85-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR