3,5-Dimethylisoxazole-4-boronic acid pinacol ester 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)isoxazole, 3,5-dimethyl-4-(tetramethyl-1,3,2-dioxaborolan-2-yl)-1,2-oxazole

A100656-100G A100656-10G A100656-25G A100656-1G A100656-5G
103795-554EA 9 USD
103795-554 103821-250 76765-120 76765-122 76765-124 76765-126 77296-786 77296-788
3,5-Dimethylisoxazole-4-boronic acid pinacol ester 98%
3,5-Dimethylisoxazole-4-boronic acid pinacol ester

Precaución: for Research Use Only

Formula: C₁₁H₁₈BNO₃
Boiling Pt: 324.7 °C (760 mm Hg)
Melting Pt: 95…99 °C
Flash Pt: 150.2 °C
Storage Temperature: Ambient
MDL Number: MFCD05863910
CAS Number: 832114-00-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR