3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 98%

Supplier: Ambeed
Warning

A377252-100MG A377252-10G A377252-250MG A377252-1G A377252-25G A377252-5G A377252-100G
103826-606EA 9 USD
103826-606 103837-634 76773-534 76773-536 76773-538 77306-896 77306-898 77306-900 77594-134
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde 98%
3,5-Dimethyl-1H-pyrrole-2-carbaldehyde

Precaución: for Research Use Only

Formula: C7H9NO
MW: 123.16 g/mol
Melting Pt: 95…99 °C
Flash Pt: 103.2 °C
Storage Temperature: Ambient
MDL Number: MFCD00111522
CAS Number: 2199-58-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR