1-Ethynyl-3,5-dimethoxybenzene 97%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Dimethoxyphenylacetylene
A160789-100G
A160789-10G
A160789-250MG
A160789-1G
A160789-25G
A160789-5G
103805-420EA
16
USD
103805-420
103843-654
77235-834
77235-836
77235-838
77319-602
77319-604
77319-606
1-Ethynyl-3,5-dimethoxybenzene 97%
1-Ethynyl-3,5-dimethoxybenzene
Precaución: for Research Use Only
Formula:
C₁₀H₁₀O₂ MW: 162.19 g/mol Boiling Pt: 264 °C (760 mm Hg) Melting Pt: 44…48 °C Flash Pt: 110 °C Storage Temperature: Ambient |
MDL Number:
MFCD03839985 CAS Number: 171290-52-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...