3,5-Dimethoxybenzylamine 97%

Supplier: Ambeed
Danger

Synonyms: 3,5-Dimethoxybenzyl amine

A110697-100G A110697-10G A110697-25G A110697-1G A110697-5G
103797-708EA 14 USD
103797-708 103823-516 77245-674 77245-676 77245-678 77310-318 77310-320
3,5-Dimethoxybenzylamine 97%
3,5-Dimethoxybenzylamine

Precaución: for Research Use Only

Formula: C₉H₁₃NO₂
MW: 167.21 g/mol
Boiling Pt: 325.7 °C (760 mm Hg)
Melting Pt: 35…37 °C
Flash Pt: 113 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00052813
CAS Number: 34967-24-3
UN: 3259
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR