3,5-Dimethoxyphenylboronic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethoxybenzeneboronic acid

A154620-100G A154620-10G A154620-1G A154620-25G A154620-5G
103804-666EA 9 USD
103804-666 103842-856 77190-888 77190-890 77300-068 77318-590 77318-592
3,5-Dimethoxyphenylboronic acid 98%
3,5-Dimethoxyphenylboronic acid

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₈H₁₁BO₄
MW: 181.98 g/mol
Boiling Pt: 371.6 °C (760 mm Hg)
Melting Pt: 205…207 °C
Flash Pt: 178.5 °C
Storage Temperature: Ambient
MDL Number: MFCD03095127
CAS Number: 192182-54-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR