3,5-Dimethoxyaniline 97%

Supplier: Ambeed
Warning

A153614-100G A153614-10G A153614-500G A153614-25G A153614-5G
103842-710EA 11 USD
103842-710 77190-882 77190-884 77190-886 77300-014 77300-016
3,5-Dimethoxyaniline 97%
3,5-Dimethoxyaniline

Precaución: Keep in a dark place, sealed in dry, room temperature

Formula: C₈H₁₁NO₂
Boiling Pt: 178 °C (760 mm Hg)
Melting Pt: 54…57 °C
Flash Pt: 135 °C
Storage Temperature: Ambient
MDL Number: MFCD00008392
CAS Number: 10272-07-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR