Sinapic acid 98%

Supplier: Ambeed
Warning

Synonyms: 3,5-Dimethoxy-4-hydroxycinnamic acid, 3',5'-Dimethoxy-4'-hydroxycinnamic acid, 4-Hydroxy-3,5-dimethoxycinnamic acid

A260988-100G A260988-100MG A260988-10G A260988-1G A260988-25G A260988-5G
103812-512EA 9 USD
103812-512 103852-400 76771-862 76771-864 76771-866 77091-994 77091-996 77091-998
Sinapic acid 98%
Sinapic acid

Precaución: for Research Use Only

Formula: C₁₁H₁₂O₅
MW: 224.21 g/mol
Boiling Pt: 403.4 °C (760 mm Hg)
Melting Pt: 191…193 °C
Flash Pt: 158.6 °C
Storage Temperature: Ambient
MDL Number: MFCD00004401
CAS Number: 530-59-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR