3,5-Diiodo-L-thyronine 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
3,5-Diiodo-L-thyronine, (S)-2-Amino-3-(4-(4-hydroxyphenoxy)-3,5-diiodophenyl)propanoic acid
A258880-1G
A258880-250MG
103812-404EA
46
USD
103812-404
103852-282
77091-812
77601-086
3,5-Diiodo-L-thyronine 95%
3,5-Diiodo-L-thyronine
Precaución: for Research Use Only
Formula:
C₁₅H₁₃I₂NO₄ MW: 525.08 g/mol Boiling Pt: 559.4 °C (760 mm Hg) Melting Pt: 255…260 °C Flash Pt: 292.1 °C Storage Temperature: Freezer |
MDL Number:
MFCD00064987 CAS Number: 1041-01-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...