3,5-Diiodobenzoic acid 97%

Supplier: Ambeed
Warning

A106620-10G A106620-250MG A106620-5G
103797-052EA 99 USD
103797-052 103822-806 77093-762 77093-764 77309-662
3,5-Diiodobenzoic acid 97%
3,5-Diiodobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₄I₂O₂
Boiling Pt: 424.2 °C (760 mm Hg)
Melting Pt: >230 °C
Flash Pt: 210.4 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00463860
CAS Number: 19094-48-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR