3,5-Dihydroxybenzaldehyde 98%

Supplier: Ambeed
Warning

A234445-1G A234445-5G A234445-10G A234445-25G A234445-100G
103810-950EA 15 USD
103810-950 103850-706 77089-692 77089-694 77140-132 77303-976 77615-930
3,5-Dihydroxybenzaldehyde 98%
3,5-Dihydroxybenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₆O₃
MW: 138.12 g/mol
Boiling Pt: 324.3 °C (760 mm Hg)
Melting Pt: 153…158 °C
Storage Temperature: Ambient
MDL Number: MFCD00016611
CAS Number: 26153-38-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR