3,5-Difluoropyridine 97%

Supplier: Ambeed
Danger

A110488-10G A110488-25G A110488-1G A110488-5G A110488-100G
103797-672EA 5 USD
103797-672 103823-478 77025-746 77025-748 77310-272 77310-274 77670-566
3,5-Difluoropyridine 97%
3,5-Difluoropyridine

Precaución: for Research Use Only

Formula: C₅H₃F₂N
MW: 115.08 g/mol
Boiling Pt: 79.4 °C (760 mm Hg)
Flash Pt: 9 °C
Storage Temperature: Refrigerator
MDL Number: MFCD03412222
CAS Number: 71902-33-5
UN: 1993
ADR: 3,Ⅱ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR