3,5-Difluoropicolinic acid 98%
Supplier: Ambeed
|
Warning
|
A107350-100G
A107350-10G
A107350-25G
A107350-1G
A107350-5G
103797-176EA
9
USD
103797-176
103822-944
77270-646
77270-648
77270-650
77283-580
77309-794
3,5-Difluoropicolinic acid 98%
3,5-Difluoropicolinic acid
Precaución: for Research Use Only
Formula:
C₆H₃F₂NO₂ Boiling Pt: 240.6 °C (760 mm Hg) Melting Pt: 202…204 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664124 CAS Number: 745784-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...