3,5-Difluorophenylboronic acid 98%

Supplier: Ambeed
Warning

A166015-100G A166015-10G A166015-500G A166015-1G A166015-25G A166015-5G
103806-010EA 7 USD
103806-010 103844-266 77096-302 77096-304 77096-306 77300-804 77320-490 77320-492
3,5-Difluorophenylboronic acid 98%
3,5-Difluorophenylboronic acid

Precaución: for Research Use Only

Formula: C₆H₅BF₂O₂
Boiling Pt: 266.9 °C (760 mm Hg)
Melting Pt: 210…217 °C
Flash Pt: 115.2 °C
Storage Temperature: Ambient
MDL Number: MFCD01318138
CAS Number: 156545-07-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR