3,5-Difluorophenol 98%

Supplier: Ambeed
Danger

A261289-25G A261289-5G A261289-100G A261289-10G A261289-1G
103852-412EA 9 USD
103852-412 77092-012 77092-014 77289-222 77289-224 77289-226
3,5-Difluorophenol 98%
3,5-Difluorophenol

Precaución: for Research Use Only

Formula: C₆H₄F₂O
MW: 130.09 g/mol
Boiling Pt: 175.4 °C (760 mm Hg)
Melting Pt: 54…57 °C
Flash Pt: 71 °C
Storage Temperature: Ambient
MDL Number: MFCD00002255
CAS Number: 2713-34-0
UN: 2925
ADR: 4.1,8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR