3,5-Difluoroisonicotinic acid 97%

Supplier: Ambeed
Warning

A138136-10G A138136-250MG A138136-25G A138136-1G A138136-5G
103802-560EA 19 USD
103802-560 76782-762 76782-764 76782-766 77298-958 77298-960
3,5-Difluoroisonicotinic acid 97%
3,5-Difluoroisonicotinic acid

Precaución: for Research Use Only

Formula: C₆H₃F₂NO₂
MW: 159.0900 g/mol
Boiling Pt: 317 °C (760 mm Hg)
Flash Pt: 146 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04972829
CAS Number: 903522-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR