3,5-Difluorobenzylamine 98%

Supplier: Ambeed
Danger

A156187-1G A156187-5G A156187-25G
103804-844EA 5 USD
103804-844 103843-048 77300-162 77300-164 77318-800
3,5-Difluorobenzylamine 98%
3,5-Difluorobenzylamine

Precaución: for Research Use Only

Formula: C₇H₇F₂N
MW: 143.14 g/mol
Boiling Pt: 184 °C (760 mm Hg)
Flash Pt: 74 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00061244
CAS Number: 90390-27-5
UN: 2735
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR