3,5-Difluorobenzaldehyde 98%

Supplier: Ambeed
Danger

A292227-100G A292227-500G A292227-10G
103835-072EA 5 USD
103835-072 77141-354 77141-356 77550-774
3,5-Difluorobenzaldehyde 98%
3,5-Difluorobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₄F₂O
Boiling Pt: 166.5 °C (760 mm Hg)
Melting Pt: 17 °C
Flash Pt: 56 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00010329
CAS Number: 32085-88-4
UN: 1989
ADR: 3,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR