3,5-Difluoro-4-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A154984-1G A154984-25G A154984-250MG
103804-704EA 20 USD
103804-704 103842-898 77300-096 77318-632 77881-215
3,5-Difluoro-4-nitrobenzonitrile 98%
3,5-Difluoro-4-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C7H2F2N2O2
Storage Temperature: Ambient
MDL Number: MFCD13193247
CAS Number: 1123172-88-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR