3,5-Difluoro-4-nitrobenzoic acid 95%

Supplier: Ambeed
Warning

A229820-250MG A229820-1G
103819-498EA 58 USD
103819-498 103850-400 77013-484 77089-250
3,5-Difluoro-4-nitrobenzoic acid 95%
3,5-Difluoro-4-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃F₂NO₄
Storage Temperature: Ambient
MDL Number: MFCD11867800
CAS Number: 1131580-60-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR