3,5-Difluoro-4-methoxyaniline 98%

Supplier: Ambeed
Warning

A261473-250MG A261473-1G A261473-25G A261473-10G
103812-524EA 20 USD
103812-524 103852-414 76692-708 77092-016 77092-018 77626-366
3,5-Difluoro-4-methoxyaniline 98%
3,5-Difluoro-4-methoxyaniline

Precaución: for Research Use Only

Formula: C₇H₇F₂NO
MW: 159.14 g/mol
Boiling Pt: 249.7 °C (760 mm Hg)
Melting Pt: 77…80 °C
Flash Pt: 104.8 °C
Storage Temperature: Refrigerator
MDL Number: MFCD04115910
CAS Number: 363-47-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR